EMAIL THIS PAGE TO A FRIEND
Iranian journal of parasitology
Species Identification and Molecular Typing of
PMID 28127360
Abstract
Leishmaniasis is a sand fly-borne disease caused by the protozoan parasites belonging to the genus A total of 130 Giemsa-Stained slides from suspected Cutaneous leishmaniasis (CL) patients were examined under a light microscope at ×1000. DNA from each slide was extracted PCR method was undertaken with HSP70 genes and the PCR products were digested with a restriction enzyme HaeIII (BsuR1). The study was conducted in the laboratory of Zahedan University of Medical Sciences in the Sistan and Baluchestan Province, southeastern Iran in 2015. From 130 suspected samples, 59 (45.3%) were positive by the microscopic examination, meanwhile 64 (49.2%) were positive by PCR-RFLP, PCR-RFLP is a valuable technique for distinguish of