EMAIL THIS PAGE TO A FRIEND
Analytical methods : advancing methods and applications
Multiplexed detection of pathogen-specific DNA using engineered zinc finger proteins without target amplification.
PMID 28127406
Abstract
Multiplexed detection of pathogen-specific DNA sequences in a simple and reliable way is in great demand for clinical and biomedical applications. However, there is still a lack of available DNA detection methods that are simple and pathogen-selective for point-of-care (POC) testing. Here, we report a novel zinc finger protein (ZFP)-based chemiluminescent method for direct detection of pathogenic double-stranded DNA (dsDNA) in a multiplexed platform. ZFPs are custom-designed to identify unique pathogenic DNA sequences. ZFP-based chemiluminescent detection of dsDNA provides sufficient sensitivity (≤50 fmol) and high specificity without target DNA amplification. Our study addresses the potential of developing a simple and selective pathogen detection method in a multiplexed fashion needed for POC application.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63423
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
C12H22O11