EMAIL THIS PAGE TO A FRIEND
Ecotoxicology and environmental safety
Prediction of biodegradability of aromatics in water using QSAR modeling.
PMID 28129599
Abstract
The study was aimed at developing models for predicting the biodegradability of aromatic water pollutants. For that purpose, 36 single-benzene ring compounds, with different type, number and position of substituents, were used. The biodegradability was estimated according to the ratio of the biochemical (BOD
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
29580
cis,cis-1,5-Cyclooctadiene, ≥98.0% (GC)
C8H12