Journal of chromatography. A
Single-step reinforced microextraction of polycyclic aromatic hydrocarbons from soil samples using an inside needle capillary adsorption trap with electropolymerized aniline/multi-walled carbon nanotube sorbent.
PMID 28129935
A polyaniline/multi-wall carbon nanotubes (PANI/MWCNT) composite was electrodeposited on the interior surface of a platinized stainless steel capillary needle and used to prepare an inside needle capillary adsorption trap (INCAT) device. The platinization expanded the interior adsorbing surface of the needle and made it more porous and cohesive for nanocomposite film. The nanocomposite was characterized using scanning electron microscopy (SEM) and Fourier transform infrared spectroscopy (FT-IR). The fabricated INCAT was fixed into a cooling capsule to fabricate a cooling-assisted INCAT (CA-INCAT) system. The CA-INCAT device was used to extract polycyclic aromatic hydrocarbons (PAHs) from solid samples followed by gas chromatography-flame ionization detection (GC-FID) determination. To obtain the best extraction efficiency, the important experimental variables were studied and optimized. Under the optimal conditions, the limits of detection (LODs) for the studied PAHs were in the range of 0.002-0.02ngg(-1). Linear dynamic ranges (LDRs) for the calibration curves were found to be 0.1-30,000ngg(-1). Relative standard deviations (RSDs%) for six replicated analysis of 1ngg(-1) PAHs were obtained 7.7-11%. The CA-INCAT-GC-FID method was successfully applied for the extraction and determination of PAHs in contaminated soil samples. The results were in agreement with those obtained by a validated ultrasound-assisted solvent extraction (UA-SE) method.
Related Materials
Product #
Molecular Formula
Add to Cart
Aniline, ACS reagent, ≥99.5%