EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
Transcriptional determinants of tolerogenic and immunogenic states during dendritic cell maturation.
PMID 28130292
Abstract
Dendritic cells (DCs) promote either tolerogenic or immunogenic T cell responses, the latter upon sensing microbes. Using an in vitro system, we analyzed transcriptional determinants that enable mature DCs to direct these opposing T cell outcomes. In the absence of microbial products, the transcription factor interferon regulatory factor 4 (IRF4) promotes regulatory T cell (T