EMAIL THIS PAGE TO A FRIEND
Metabolic brain disease
Investigation of the motor system in two siblings with Canavan's disease: a combined transcranial magnetic stimulation (TMS) - diffusion tensor imaging (DTI) study.
PMID 28130616
Abstract
Canavan's disease (CD) is a hereditary leukodystrophy caused by mutations in the aspartoacylase gene (ASPA), leading to spongiform degeneration of the white matter and severe impairment of psychomotor development. We present the cases of two non-Jewish sisters with CD that have a milder and protracted clinical course compared to typical CD. MRI imaging revealed bilateral high-signal-intensity areas in the thalami and the internal capsule and MR spectroscopy showed typical findings for CD (a marked increase in N-acetylaspartate (NAA) levels). FA values of the right and left corticospinal tracts at the level of the posterior limb of the internal capsule, and the centrum semiovale were found to be significantly reduced compared to healthy controls. From a neurophysiological point of view, the peripheral motor system was normal. In contrast, cortical stimulation at maximal intensity failed to elicit facilitated or resting MEPs and silent periods (SPs) in upper and lower limbs, providing evidence for significant upper motor pathway dysfunction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
719536
3,3′-Diiodo-L-thyronine (T2), 98% (CP)
C15H13I2NO4