EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Impedimetric nanostructured genosensor for detection of schistosomiasis in cerebrospinal fluid and serum samples.
PMID 28131054
Abstract
Schistosomiasis is a neglected disease closely related to the low levels of social development and a serious public health problem. In this work, we performed an electrochemical detection of Schistosoma mansoni DNA with a self-assembled monolayer of mercaptobenzoic acid (MBA) immobilizing nanostructures composed of gold nanoparticles (AuNPs) and magnetite nanoparticles (Fe
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T33200
Thiosalicylic acid, 97%
C7H6O2S