EMAIL THIS PAGE TO A FRIEND
Saudi medical journal
Gender differences in risk profile and outcome of Middle Eastern patients undergoing percutaneous coronary intervention.
PMID 28133687
Abstract
To determine the gender differences in cardiovascular risk profile and outcomes among patients undergoing percutaneous coronary intervention (PCI). Methods: In a prospective multicenter study of consecutive Middle Eastern patients managed with PCI from January 2013 to February 2014 in 12 tertiary care centers in Amman and Irbid, Jordan. Clinical and coronary angiographic features, and major cardiovascular events were assessed for both genders from hospital stay to 1 year. Results: Women comprised 20.6% of 2426 enrolled patients, were older (mean age 62.9 years versus 57.2 years), had higher prevalence of hypertension (81% versus 57%), diabetes (66% versus 44%), dyslipidemia (58% versus 46%), and obesity (44% versus 25%) compared with men, p less than 0.001. The PCI for ST-segment elevation myocardial infarction was indicated for fewer women than men (23% versus 33%; p=0.001). Prevalence of single or multi-vessel coronary artery disease was similar in women and men. More women than men had major bleeding during hospitalization (2.2% versus 0.6%; p=0.003) and at one year (2.5% versus 0.9%; p=0.007). There were no significant differences between women and men in mortality (3.1% versus 1.7%) or stent thrombosis (2.1% versus 1.8%) at 1 year. Conclusion: Middle Eastern women undergoing PCI had worse baseline risk profile compared with men.Except for major bleeding, no gender differences in the incidence of major adverse cardiovascular events were demonstrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5942
DAPT, ≥98% (HPLC), solid
C23H26F2N2O4