EMAIL THIS PAGE TO A FRIEND
Scientific reports
Hydrocracking of Jatropha Oil over non-sulfided PTA-NiMo/ZSM-5 Catalyst.
PMID 28134313
Abstract
The PTA-NiMo/ZSM-5 catalyst impregnated with phosphotungstic acid (PTA) was designed for the transformation of Jatropha oil into benzene, toluene, and xylenes (BTX) aromatics. The produced catalyst was characterized by N
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
469548
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
C8H11F3O2