EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Colorimetric detection of endogenous hydrogen sulfide production in living cells.
PMID 28135697
Abstract
Hydrogen sulfide (H
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69453
L-Homocysteine, ≥98.0% (NT)
C4H9NO2S