EMAIL THIS PAGE TO A FRIEND
Respiratory physiology & neurobiology
Effects on breathing of agonists to μ-opioid or GABA
PMID 28137700
Abstract
Pulmonary ventilation (V̇
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7384
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
C26H35N5O6