Journal of applied toxicology : JAT
Lethal and sublethal effects of aniline and chlorinated anilines on zebrafish embryos and larvae.
PMID 28138964
Environmental risk assessments show increased attention to the sublethal effects of chemicals on aquatic organisms. The Organization for Economic Cooperation and Development (OECD) established the "Fish, Short-term Toxicity Test on Embryo and Sac-fry Stages" (OECD test 212) to predict lethal effects. It is still unclear, however, whether this test can predict sublethal effects. Although their sublethal effects are still unknown, chlorinated anilines are widely used in various fields. The purpose of this study, therefore, is to investigate sublethal effects of chlorinated anilines using OECD test 212 with zebrafish, and to examine the correlation of several sublethal effects between embryo and larval stages. Embryos were exposed to aniline and nine chlorinated anilines until 8 days post-fertilization. A delayed lethal effect was observed from three of the 10 anilines tested. In the control group, the swim bladder inflated after hatching, but there was no swim-bladder inflation after exposure to the chlorinated anilines. Fertilized eggs exposed to lower concentrations of test chemicals showed effects during embryogenesis that did not affect mortality rates, such as changes in body curvature and edema. Our results show that chlorinated anilines induce not only lethal effects but also a variety of sublethal effects. Moreover, a detailed estimate of these effects requires study during both embryonic and larval stages. OECD test 212 may therefore prove useful as a method for screening chemicals for lethal and sublethal effects. Copyright © 2017 John Wiley & Sons, Ltd.
Related Materials
Product #
Molecular Formula
Add to Cart
4-Ethylaniline, 98%