EMAIL THIS PAGE TO A FRIEND
Luminescence : the journal of biological and chemical luminescence
Polymer composite fluorescent hydrogel film based on nitrogen-doped carbon dots and their application in the detection of Hg
PMID 28139046
Abstract
A simple microwave-assisted solvothermal method was used to prepare fluorescent nitrogen-doped carbon dots (N-CDs) with high fluorescence quantum yield (79.63%) using citric acid and N-(2-hydroxyethyl)ethylenediamine as starting materials. The PVAm-g-N-CDs grafted products were synthesized by amide bond formation between the carboxylic groups of N-CDs and amine groups of polyvinylamine (PVAm). Fluorescent hydrogel films (PVAm-g-N-CDs/PAM) were synthesized by interpenetration polymer network polymerization of PVAm-g-N-CDs and acrylamide (AM). When used for ion detection, we found that the fluorescence of the hydrogel films was clearly quenched by addition of Hg