EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
A minimal-invasive method for systemic bio-monitoring of the environmental pollutant phenanthrene in humans: Thermal extraction and gas chromatography - mass spectrometry from 1 mL capillary blood.
PMID 28139229
Abstract
Phenanthrene is present in numerous environmental media and serves as a model substrate for the biomonitoring of polycyclic aromatic hydrocarbon (PAH). PAH exposure studies are commonly focused on urinary metabolites, concentrations of which are dependent on absorption, biotransformation and excretion. Monitoring of unmetabolized PAHs in blood would allow more reliable exposure assessment, but requires invasive sampling and extensive sample preparation. We describe the analysis of phenanthrene in 1μL capillary blood using thermal extraction (TE) combined with gas chromatography - mass spectrometry (GC-MS). Less invasive sampling of 1μL capillary blood does not require the assistance of medical staff. Compared to previous studies, analysis time was improved significantly by TE due to minimization of sample preparation steps. The evaluate method was applied successfully to the monitoring of phenanthrene blood levels. This is the first report presenting the pharmacokinetics of unmetabolized PAHs in human.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
217042
4,4,4-Trifluoro-1-phenyl-1,3-butanedione, 99%
C10H7F3O2