EMAIL THIS PAGE TO A FRIEND
BMC complementary and alternative medicine
Chemical composition, vasorelaxant, antioxidant and antiplatelet effects of essential oil of Artemisia campestris L. from Oriental Morocco.
PMID 28143473
Abstract
Artemisia campestris L. (Asteraceae) is a medicinal herb traditionally used to treat hypertension and many other diseases. Hence, this study is aimed to analyze the essential oil of A. campestris L (AcEO) and to investigate the antiplatelet, antioxidant effects and the mechanisms of its vasorelaxant effect. The chemical composition of AcEO was elucidated using GC/MS analysis. Then, the antioxidant effect was tested on DPPH radical scavenging and on the prevention of β-carotene bleaching. The antiplatelet effect was performed on the presence of the platelet agonists: thrombin and ADP. The mechanism of action of the vasorelaxant effect was studied by using the cellular blockers specified to explore the involvement of NO/GC pathway and in the presence of calcium channels blockers and potassium channels blockers. AcEO is predominated by the volatiles: spathulenol, ß-eudesmol and p-cymene. The maximal antioxidant effect was obtained with the dose 2xa0mg/ml of AcEO. The dose 1xa0mg/ml of AcEO showed a maximum antiplatelet effect of, respectively 49.73% ±9.54 and 48.20% ±8.49 on thrombin and ADP. The vasorelaxation seems not to be mediated via NOS/GC pathway neither via the potassium channels. However, pretreatment with calcium channels blockers attenuated this effect, suggesting that the vasorelaxation is mediated via inhibition of L-type Ca(2+) channels and the activation of SERCA pumps of reticulum plasma. This study confirms the antioxidant, antiplatelet and vasorelaxant effects of A.campestris L essential oil. However, the antihypertensive use of this oil should be further confirmed by the chemical fractionation and subsequent bio-guided assays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3930
Calmidazolium chloride, solid
C31H23Cl7N2O