EMAIL THIS PAGE TO A FRIEND
Molecular neurodegeneration
APOE Genotype Differentially Modulates Effects of Anti-Aβ, Passive Immunization in APP Transgenic Mice.
PMID 28143566
Abstract
APOE genotype is the foremost genetic factor modulating β-amyloid (Aβ) deposition and risk of sporadic Alzheimer's disease (AD). Here we investigated how APOE genotype influences response to anti-Aβ immunotherapy. APP Anti-Aβ immunization decreased both the load of fibrillar plaques and the load of Aβ immunopositive plaques in mice of all APOE backgrounds. Although the relative reduction in parenchymal Aβ plaque load was comparable across all APOE genotypes, APP/ε4 mice showed the greatest reduction in the absolute Aβ plaque load values, given their highest baseline. The immunization stimulated phagocytic activation of microglia, which magnitude adjusted for the post-treatment plaque load was the greatest in APP/ε4 mice implying association between the ε4 allele and impaired Aβ phagocytosis. Perivascular hemosiderin deposits reflecting ensued microhemorrhages were associated with vascular Aβ (VAβ) and ubiquitously present in control mice of all APOE genotypes, although in APP/ε3 mice their incidence was the lowest. Anti-Aβ immunization significantly reduced VAβ burden but increased the number of hemosiderin deposits across all APOE genotypes with the strongest and the weakest effect in APP/ε2 and APP/ε3 mice, respectively. Our studies indicate that APOE genotype differentially modulates microglia activation and Aβ plaque load reduction during anti-Aβ immunotherapy. The APOE ε3 allele shows strong protective effect against immunotherapy associated microhemorrhages; while, conversely, the APOE ε2 allele increases risk thereof.