EMAIL THIS PAGE TO A FRIEND
Beilstein journal of organic chemistry
Versatile synthesis of end-reactive polyrotaxanes applicable to fabrication of supramolecular biomaterials.
PMID 28144361
Abstract
Cyclodextrin (CD)-threaded polyrotaxanes (PRXs) with reactive functional groups at the terminals of the axle polymers are attractive candidates for the design of supramolecular materials. Herein, we describe a novel and simple synthetic method for end-reactive PRXs using bis(2-amino-3-phenylpropyl) poly(ethylene glycol) (PEG-Ph-NH