EMAIL THIS PAGE TO A FRIEND
Zoological science
Upregulation of Juvenile Hormone Titers in Female Drosophila melanogaster Through Mating Experiences and Host Food Occupied by Eggs and Larvae.
PMID 28148219
Abstract
Juvenile hormone (JH) plays a crucial role in the determination of developmental timing in insects. In Drosophila melanogaster, reports indicate that JH titers are the highest immediately following eclosion and that the mating experience increases the titers in females. However, the titers have not been successively measured for an extended period of time after eclosion. This study reveals that JH titers are increased after eclosion in virgin females when supplied with food that is occupied by eggs and larvae as well as in mated females. With the presence of eggs and larvae, food induced the virgin females to lay unfertilized eggs. When combined with previous work indicating that females are attracted to such food where they prefer to lay eggs, these results suggest that flies can prepare themselves to lay eggs by changing the titers of JH under the presence of growing larvae, ensuring that the food is an appropriate place to oviposit.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34343
Fenoxycarb, PESTANAL®, analytical standard
C17H19NO4
J2000
Juvenile hormone III, ≥65%
C16H26O3