EMAIL THIS PAGE TO A FRIEND
ACS central science
CYP450 Enzymes Effect Oxygen-Dependent Reduction of Azide-Based Fluorogenic Dyes.
PMID 28149949
Abstract
Azide-containing compounds have broad utility in organic synthesis and chemical biology. Their use as powerful tools for the labeling of biological systems
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU099041 MISSION® esiRNA, esiRNA human CBS (esiRNA1)