EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Effects of the Hofmeister series of sodium salts on the solvent properties of water.
PMID 28150000
Abstract
The solvent features of water (solvent dipolarity/polarizability, π*, hydrogen bond donor acidity, α, and hydrogen bond acceptor basicity, β) were examined in aqueous solutions of Na
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103543
4-Nitroanisole, 97%
C7H7NO3