EMAIL THIS PAGE TO A FRIEND
Scientific reports
Legacy of contaminant N sources to the NO
PMID 28150819
Abstract
Nitrate content of surface waters results from complex mixing of multiple sources, whose signatures can be modified through N reactions occurring within the different compartments of the whole catchment. Despite this complexity, the determination of nitrate origin is the first and crucial step for water resource preservation. Here, for the first time, we combined at the catchment scale stable isotopic tracers (δ
Related Materials