EMAIL THIS PAGE TO A FRIEND
Talanta
Determination of trace Se(IV) by anodic stripping voltammetry following double deposition and stripping steps.
PMID 28153268
Abstract
A procedure of Se(IV) determination by anodic stripping voltammetry using two deposition and stripping steps at gold electrodes was proposed. A well-defined stripping peak of selenium was obtained at potential 0.9 V. The optimization of parameters influencing the selenium peak current including both deposition and stripping steps was performed. A linear relationship was observed between the Se(IV) peak current and its concentration in the range from 5×10