EMAIL THIS PAGE TO A FRIEND
Talanta
Development simple and sensitive voltammetric procedure for ultra-trace determination of U(VI).
PMID 28153285
Abstract
A simple and sensitive adsorptive stripping voltammetric procedure for the determination of trace concentration of U(VI) in natural water samples was developed. In order to remove surface active compounds from water sample solutions, a TiO