EMAIL THIS PAGE TO A FRIEND
International journal of pharmaceutics
In vitro permeation and disposition of niacinamide in silicone and porcine skin of skin barrier-mimetic formulations.
PMID 28153652
Abstract
Niacinamide (NIA) is an amide form of vitamin B3 which is used in cosmetic formulations to improve various skin conditions and it has also been shown to increase stratum corneum thickness following repeated application. In this study, three doses (5, 20 and 50μL per cm
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
469319
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
C8H24O2Si3