EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
CdTe quantum dot-based fluorescent probes for selective detection of Hg (II): The effect of particle size.
PMID 28153811
Abstract
Mercury ions-induced fluorescence quenching properties of CdTe quantum dots (QDs) have been studied using the fluorescence spectroscopic techniques. By using the hydrothermal method, the CdTe QDs with different particles sizes from 1.98 to 3.68nm have been prepared, and the corresponding fluorescence emission wavelength is changed from 518 to 620nm. The fluorescence of QDs is enhanced after linking Bovine serum albumin (BSA) onto the surface of the QDs. Experimental results show that the fluorescence intensity of BSA-coated CdTe QDs could be effectively quenched when Hg(2+) react with BSA-coated CdTe QDs. Interestingly, both the sensing sensitivity and selectivity of this fluorescence probe could be improved when the particle size of the QDs decreases. Thus the BSA-coated CdTe QDs with green fluorescence emission have better advantages than the BSA-coated CdTe QDs with red fluorescence for Hg(2+) detection. Interference experiment results indicate that the influence from other metal ions could be neglected in the detection, and the Hg(2+) could be specifically detected. By using this BSA-coated CdTe QDs-based fluorescence probe, the Hg(2+) could be detected with an ultra-low detection limit of nanomole level, and the linear range spans a scope from 0.001 to 1μmol/L.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3758
Thioglycolic acid, 98%
C2H4O2S