EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Epigenetic Activation of
PMID 28153853
Abstract
Since the discovery of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2625
Retinoic acid, ≥98% (HPLC), powder
C20H28O2