EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Gene polymorphism linked to increased asthma and IBD risk alters gasdermin-B structure, a sulfatide and phosphoinositide binding protein.
PMID 28154144
Abstract
The exact function of human gasdermin-B (GSDMB), which regulates differentiation and growth of epithelial cells, is yet to be elucidated. In human epidermal growth factor receptor 2 (HER2)-positive breast cancer,
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63423
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
C12H22O11