EMAIL THIS PAGE TO A FRIEND
Journal of nematology
Grafting and Paladin Pic-21 for Nematode and Weed Management in Vegetable Production.
PMID 28154429
Abstract
Two years of field trials conducted in a
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33859
Metolachlor, predominantly S-Metolachlor, PESTANAL®, analytical standard
C15H22ClNO2