EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Synthesis of OMS-2/graphite nanocomposites with enhanced activity for pollutants degradation in the presence of peroxymonosulfate.
PMID 28157637
Abstract
The octahedral molecular sieve manganese oxide (OMS-2) and graphite (Gt) composites (OMS-2/Gt) were prepared by a facile refluxing approach. The structure and morphology of the obtained materials were systematically investigated. The results reveal that OMS-2 nanofibers are uniformly dispersed on the surface of Gt, and the ratio of Mn
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
228036
OXONE®, monopersulfate compound
HKO5S · 0.5HKO4S · 0.5K2O4S
P9625
Phenazine methosulfate, ≥90% (UV)
C14H14N2O4S