EMAIL THIS PAGE TO A FRIEND
Food chemistry
Identification and quantification of ACE-inhibiting peptides in enzymatic hydrolysates of plant proteins.
PMID 28159254
Abstract
Enzymatic hydrolysis of proteins from rice, soy, pea and wheat, with both chymotrypsin and thermolysin, resulted in hydrolysates, which are efficient inhibitors of the angiotensin-converting enzyme (ACE). IC