EMAIL THIS PAGE TO A FRIEND
Food chemistry
Phytochemical composition of fractions isolated from ten Salvia species by supercritical carbon dioxide and pressurized liquid extraction methods.
PMID 28159282
Abstract
Ten Salvia species, S. amplexicaulis, S. austriaca, S. forsskaolii S. glutinosa, S. nemorosa, S. officinalis, S. pratensis, S. sclarea, S. stepposa and S. verticillata were fractionated using supercritical carbon dioxide and pressurized liquid (ethanol and water) extractions. Fifteen phytochemicals were identified using commercial standards (some other compounds were identified tentatively), 11 of them were quantified by ultra high pressure chromatography (UPLC) with quadruple and time-of-flight mass spectrometry (Q/TOF, TQ-S). Lipophilic CO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48640
Ethyl gallate, antioxidant, ≥96.0% (HPLC)
C9H10O5
17799
Rhamnetin, analytical standard
C16H12O7