EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Selective chromatographic separation of polycarbonate according to hydroxyl end-groups using a porous graphitic carbon column.
PMID 28159367
Abstract
Porous graphitic carbon (PGC) has shown unique separation efficiency in liquid chromatography for a wide range of substance classes. In the characterization of polymers PGC has particularly been used for analysis of polyolefins. Its retention mechanisms differ dramatically from those of silica-based stationary phases and therefore allow interesting applications. Due to its unprecedented retention mechanisms PGC does not only promise good separation performance for polyolefins but also for more polar polymers such as Polycarbonate (PC). In this study, we determined the critical conditions of PC on PGC using CHCl
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
752193
1,3,5-Tris(N-carbazolyl)benzene, 97%
C42H27N3