EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Heart and circulatory physiology
Cardiac Med1 deletion promotes early lethality, cardiac remodeling, and transcriptional reprogramming.
PMID 28159809
Abstract
The mediator complex, a multisubunit nuclear complex, plays an integral role in regulating gene expression by acting as a bridge between transcription factors and RNA polymerase II. Genetic deletion of mediator subunit 1 (Med1) results in embryonic lethality, due in large part to impaired cardiac development. We first established that Med1 is dynamically expressed in cardiac development and disease, with marked upregulation of Med1 in both human and murine failing hearts. To determine if Med1 deficiency protects against cardiac stress, we generated two cardiac-specific