EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and chemistry
Analysis of transcriptional responses of normalizing genes on Crassostrea brasiliana under different experimental conditions.
PMID 28160493
Abstract
Bivalves show remarkable plasticity to environmental changes and have been proposed as sentinel organisms in biomonitoring. Studies related to transcriptional analysis using quantitative real-time polymerase chain reaction (qRT-PCR) in these organisms have notably increased, imposing a need to identify and validate adequate reference genes for an accurate and reliable analysis. In the present study, 9 reference genes were selected from transcriptome data of Crassostrea brasiliana to identify their suitability as qRT-PCR normalizer genes. The transcriptional patterns were analyzed in gills of oysters under 3 different conditions: different temperatures (18, 24, or 32 °C) and phenanthrene (100 µg L