EMAIL THIS PAGE TO A FRIEND
Biomaterials
Effect of sex-hormone levels, sex, body mass index and other host factors on human craniofacial bone regeneration with bioactive tricalcium phosphate grafts.
PMID 28160669
Abstract
Little is known regarding the associations between sex-hormone levels, sex, body mass index (BMI), age, other host factors and biomaterial stimulated bone regeneration in the human craniofacial skeleton. The aim of this study was to elucidate the associations between these factors and bone formation after sinus floor augmentation procedures (SFA) utilizing a bioactive tricalcium phosphate (TCP) bone grafting material. We conducted a prospective study in a human population in which 60 male and 60 female participants underwent SFA and dental implant placement using a staged approach. BMI as well as levels of serum estradiol (E2), total testosterone (TT), and the free androgen index (FAI) were measured by radioimmunoassay and electrochemoluminescent-immunoassay. At implant placement, 6 months after SFA, bone biopsy specimens were harvested for hard tissue histology, the amount of bone formation was evaluated by histomorphometry and immunohistochemical analysis of osteogenic marker expression. The Wilcoxon rank-sum U test, Spearman correlations and linear regression analysis were used to explore the association between bone formation and BMI, hormonal and other host factors. BMI and log E2 were significantly positively associated with bone formation in male individuals (p < 0.05). Histomorphometry revealed trends toward greater bone formation and osteogenic marker expression with non-smokers compared to smokers. In male patients, higher E2 levels and higher BMI enhanced TCP stimulated craniofacial i.e. intramembranous bone repair.