EMAIL THIS PAGE TO A FRIEND
Redox biology
Mitochondrial contribution to lipofuscin formation.
PMID 28160744
Abstract
Mitochondria have been in the focus of oxidative stress and aging research for decades due to their permanent production of ROS during the oxidative phosphorylation. The hypothesis exists that mitochondria are involved in the formation of lipofuscin, an autofluorescent protein aggregate that accumulates progressively over time in lysosomes of post-mitotic and senescent cells. To investigate the influence and involvement of mitochondria in lipofuscinogenesis, we analyzed lipofuscin amounts as well as the mitochondrial function in young and senescent cells. In addition we used an aging model and Lon protease deficient HeLa cells to investigate the influence of mitochondrial degradation processes on lipofuscin formation. We were able to show that mitophagy is impaired in senescent cells resulting in an increased mitochondrial mass and superoxide formation. In addition, the inhibition of mitochondrial fission leads to increased lipofuscin formation. Moreover, we observed that Lon protease downregulation is linked to a higher lipofuscinogenesis whereas the application of the mitochondrial-targeted antioxidant mitoTEMPO is able to prevent the accumulation of this protein aggregate.