EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Hydrothermal synthesis of manganese phosphate/graphene foam composite for electrochemical supercapacitor applications.
PMID 28161504
Abstract
Manganese phosphate (Mn
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
330825
Manganese(II) acetate, 98%
C4H6MnO4