EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
SIRT3 deacetylated and increased citrate synthase activity in PD model.
PMID 28161643
Abstract
SIRT3 have been found to be neuroprotective in many neurological diseases, but its detail mechanism is only partially understood. In this study, MPP
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T5428
Tetramethylrhodamine methyl ester perchlorate, ≥95%
C25H25ClN2O7