EMAIL THIS PAGE TO A FRIEND
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP
Overexpression of carboxylesterase contributes to the attenuation of cyanotoxin microcystin-LR toxicity.
PMID 28163251
Abstract
Microcystin-LR is a hepatotoxin produced by several cyanobacteria. Its toxicity is mainly due to a inhibition of protein phosphatase, PP1 and PP2A. Previously, we used a cell line stably expressing uptake transporter for microcystin-LR, OATP1B3 (HEK293-OATP1B3 cells). In this study, to determine whether overexpression of carboxylesterase (CES), which degrades ester-group and amide-group, attenuates the cytotoxicity of microcystin-LR, we generated the HEK293-OATP1B3/CES2 double-transfected cells. HEK293-OATP1B3/CES2 cells showed high hydrolysis activity of p-nitrophenyl acetate (PNPA), which is an authentic substrate for esterase. CES activity in HEK293-OATP1B3/CES2 cells was approximately 3-fold higher than that in the HEK293-OATP1B3 cells. HEK293-OATP1B3/CES2 cells (IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33893
Microcystin-LR solution, 10 μg/mL in methanol, analytical standard
C49H74N10O12