EMAIL THIS PAGE TO A FRIEND
ChemistryOpen
Self-Supported Polypyrrole/Polyvinylsulfate Films: Electrochemical Synthesis, Characterization, and Sensing Properties of Their Redox Reactions.
PMID 28168147
Abstract
Thick films of polypyrrole/polyvinylsulfate (PPy/PVS) blends were electrogenerated on stainless-steel electrodes under potentiostatic conditions from aqueous solution. The best electropolymerization potential window was determined by cyclic voltammetry. After removing the film from the back metal, self-supported electrodes were obtained. Voltammetric, coulovoltammetric, and chronoamperometric responses from a LiClO