EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Synthesis and evaluation of new copper ketoiminate precursors for a facile and additive-free solution-based approach to nanoscale copper oxide thin films.
PMID 28170011
Abstract
Novel copper ketoiminate compounds were synthesized and for the first time applied for additive-free solution-based deposition of nanoscale copper oxide thin films. The two closely related compounds, namely the bis[4-(2-ethoxyethyl-imino)-3-pentanonato]copper, [Cu(EEKI)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
774243
1,3-Propylene sulfite, 99%
C3H6O3S