EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Monoclonal antibody defining a stage-specific mouse embryonic antigen (SSEA-1).
PMID 281705
Abstract
A monoclonal antibody derived by fusion of mouse myeloma cells with spleen cells from a mouse immunized with F9 teratocarcinoma cells is described. This antibody, which reacts with embryonal carcinoma cells of mouse and human origin and with some preimplantation stage mouse embryos, defines an embryonic stage-specific antigen. This stage-specific antigen (SSEA-1) is first detected on blastomeres of 8-cell stage embryos. Trophectodermal cells are transitorily positive; however, each cell in the inner cell mass eventually expresses this antigen.