EMAIL THIS PAGE TO A FRIEND
Chemosphere
A comparative study of ozonation, iron coated zeolite catalyzed ozonation and granular activated carbon catalyzed ozonation of humic acid.
PMID 28171838
Abstract
This study compares ozonation (O
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6875
21-Hydroxyprogesterone, ≥97% (HPLC)
C21H30O3