EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
Sampling for silver nanoparticles in aqueous media using a rotating disk electrode: evidence for selective sampling of silver nanoparticles in the presence of ionic silver.
PMID 28176261
Abstract
Amendment of a carbon paste electrode consisting of graphite and Nujol®, with a variety of organic and inorganic materials, allows direct adsorption of silver nanoparticles (AgNPs) from aqueous solution in either open or close circuit modes. The adsorbed AgNPs are detected by stripping voltammetry. Detection limits of less than 1 ppb Ag are achievable with a rotating disk system. More than one silver peak was apparent in many of the stripping voltammograms. The appearance of multiple peaks could be due to different species of silver formed upon stripping or variation in the state of aggregation or size of nanoparticles. With most of these packing materials, dissolved Ag
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
71063
Goethite, 30-63% Fe
HFeO2