EMAIL THIS PAGE TO A FRIEND
Journal of the peripheral nervous system : JPNS
Novel NTRK1 mutations associated with congenital insensitivity to pain with anhidrosis verified by functional studies.
PMID 28177573
Abstract
Congenital insensitivity to pain with anhidrosis (CIPA), also known as hereditary sensory and autonomic neuropathy type IV, features loss of pain sensation, decreased or absent sweating (anhidrosis), recurrent episodes of unexplained fever, self-mutilating behavior, and variable mental retardation. Mutations in neurotrophic receptor tyrosine kinase 1 (NTRK1) have been reported to be associated with CIPA. We identified four novel NTRK1 mutations in six Korean patients from four unrelated families. Of the four mutations, we demonstrated using a splicing assay that IVS14+3A>T causes aberrant splicing of NTRK1 mRNA, leading to introduction of a premature termination codon. An NTRK1 autophosphorylation assay showed that c.1786G>A (p.Asp596Asn) abolished autophosphorylation of NTRK1. In addition, Western blotting showed that c.704C>G (p.Ser235*) and c.2350_2363del (p.Leu784Serfs*79) blunted NTRK1 expression to undetectable levels. The four novel NTRK1 mutations we report here will expand the repertoire of NTRK1 mutations in CIPA patients, and further our understanding of CIPA pathogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9405
O-Phospho-L-tyrosine
C9H12NO6P