Journal of biomedical materials research. Part A
Student Award for Outstanding Research Winner in the Undergraduate Category for the 2017 Society for Biomaterials Annual Meeting and Exposition, April 5-8, 2017, Minneapolis, Minnesota: Development and characterization of stimuli-responsive hydrogel microcarriers for oral protein delivery.
PMID 28177593
A family of pH-responsive terpolymers composed of methacrylic acid (MAA), N-vinyl pyrrolidone (NVP), and poly(ethylene glycol) monomethylether monomethacrylate (PEGMMA) have been developed and evaluated for their pH-responsive swelling behavior, protein-loading capabilities, and cytocompatibility. These terpolymer hydrogels, designated as P((MAA-co-NVP)-g-EG), were synthesized with varying PEG chain lengths and monomer feed ratios. The incorporation of MAA into the terpolymer structure was quantified with potentiometric titration. Equilibrium and dynamic swelling studies confirmed the pH-responsive behavior of the hydrogel, with the system remaining collapsed/complexed in acidic pH conditions and swollen/decomplexed in neutral pH conditions. The ability of the hydrogels to partition protein into the swollen network was assessed for two model proteins of varying molecular weight: insulin and porcine growth hormone. Finally, the cytocompatibility of the hydrogels in the presence of two model intestinal cell lines was investigated and confirmed minimal cytotoxicity at and below 2.5 mg/mL. The developed P((MAA-co-NVP)-g-EG) hydrogels exhibit unique properties that could potentially be utilized for drug delivery and separation applications. © 2017 Wiley Periodicals, Inc. J Biomed Mater Res Part A: 105A: 1243-1251, 2017.
Related Materials
Product #
Molecular Formula
Add to Cart
3,3-Dimethylglutaric acid, 98%