EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
A novel LCMSMS method for quantitative measurement of short-chain fatty acids in human stool derivatized with
PMID 28178633
Abstract
A novel liquid chromatography tandem mass spectrometry (LCMSMS) method for the quantitative measurement of gut microbial-derived short-chain fatty acids (SCFAs) in human infant stool has been developed and validated. Baseline chromatographic resolution was achieved for 12 SCFAs (acetic, butyric, caproic, 2,2-dimethylbutyric, 2-ethylbutyric, isobutyric, isovaleric, 2-methylbutyric, 4-methylvaleric, propionic, pivalic and valeric acids) within an analysis time of 15min. A novel sequential derivatization of endogenous and spiked SCFAs in stool via
Related Materials