EMAIL THIS PAGE TO A FRIEND
Anticancer research
HIF1α in Tumorigenesis of Adenoid Cystic Carcinoma.
PMID 28179306
Abstract
Tumor hypoxia induces hypoxia-inducible factor-1α (HIF1α), which can influence tumorigenesis and metastasis. We evaluated the expression of HIF1α and the effect of HIF1α inhibitors in adenoid cystic carcinoma (ACC). HIF1α expression was demonstrated in ACC cell lines (ACC2 and ACCM). The effect of HIF1α inhibitors was evaluated. A systemic metastasis model was developed. The number of metastatic pulmonary nodules were analyzed. The ACCM cell line demonstrated greater HIF1α expression and invasion than ACC2. The expression of HIF1α and invasion of ACC cells were blocked by HIF1α siRNA. HIF1α inhibitors 17-N-allylamino-17-demethoxygeldanamycin (17AAG) and echinomycin inhibited cell invasion. 17AAG inhibited metastasis in the animal model, although not statistically significantly. HIF1α siRNA and 17AAG and echinomycin blocked invasion by ACC2 and ACCM cells. 17AAG exhibited therapeutic potential for inhibition of metastasis. Our results provide positive evidence that HIF1α is a promising research pathway for therapy of ACC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1043
KC7F2, ≥98% (HPLC)
C16H16Cl4N2O4S4