EMAIL THIS PAGE TO A FRIEND
Anticancer research
Down-regulation of RASA1 Is Associated with Poor Prognosis in Human Hepatocellular Carcinoma.
PMID 28179330
Abstract
RASA1 (p120RasGAP), encodes Ras GTPase-activating protein 1 and, is a potent tumor suppressor gene that is frequently inactivated in several human cancer types. However, its precise role in hepatocellular carcinoma (HCC) has been blurred. We hypothesized that RASA1 plays a crucial role in tumor pathogenesis and progression of HCC. RASA1 expression levels were analyzed in 226 cases of HCC by immunohistochemistry. It was found that 38.68% (41/106) of the high-grade HCC samples and 54.17% (65/120) of the low-grade HCC samples expressed RASA1 protein. The difference between RASA1 expression in high-grade and low-grade HCC was statistically significant (p=0.02). Additionally, RASA1 high expression was inversely associated with larger tumor size (p<0.001). Although RASA1 is known as a tumor suppressor, its role in overall survival (OS) in HCC is unclear. Kaplan-Meier survival analysis showed that patients with low level of RASA1 expression correlated with a significantly poorer survival compared to those with high level of RASA1 expression. These data support that RASA1 could serve as an independent prognostic marker for HCC patients.