EMAIL THIS PAGE TO A FRIEND
Cell death & disease
TAK1 regulates resident macrophages by protecting lysosomal integrity.
PMID 28182011
Abstract
Hematopoietic cell survival and death is critical for development of a functional immune system. Here, we report that a protein kinase, TAK1, is selectively required for resident macrophage integrity during embryogenesis. Hematopoietic lineage-specific deletion of Tak1 gene (Tak1